Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 27342006-27342249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL4R
Entrez Gene ID
3566
Gene Name
interleukin 4 receptor
Gene Aliases
CD124, IL-4RA, IL4RA
UniGene
Hs.513457
Gene Chromosome Location
Chr.16: 27313668-27364778 on Build GRCh38
Transcripts
NM_000418.3 NM_001257406.1 NM_001257407.1 NM_001257997.1 XM_011545825.1 XM_011545826.2 XM_011545827.2 XM_011545828.2 XM_011545829.1 XM_011545830.2 XM_011545831.1 XM_011545832.1 XM_011545833.2 XM_011545834.2 XM_017023211.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1194906 c.125C>G p.S42C Chr.16: 27342175-27342175 on Build GRCh38
7153535 c.188A>G p.Q63R Chr.16: 27342238-27342238 on Build GRCh38
4059625 c.85T>C p.L29L Chr.16: 27342135-27342135 on Build GRCh38
3108710 c.135G>A p.E45E Chr.16: 27342185-27342185 on Build GRCh38
1129007 c.108C>T p.S36S Chr.16: 27342158-27342158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17548704 Chr.16: 27342158-27342158 on Build GRCh38
rs17548697 Chr.16: 27342094-27342094 on Build GRCh38
rs200558051 Chr.16: 27342108-27342108 on Build GRCh38
rs770213018 Chr.16: 27342226-27342226 on Build GRCh38
rs776677335 Chr.16: 27342159-27342159 on Build GRCh38
rs908910299 Chr.16: 27342142-27342142 on Build GRCh38
rs55671142 Chr.16: 27342152-27342152 on Build GRCh38
rs1051167644 Chr.16: 27342183-27342183 on Build GRCh38
rs1017505097 Chr.16: 27342079-27342079 on Build GRCh38
rs191774649 Chr.16: 27342225-27342225 on Build GRCh38
rs3024544 Chr.16: 27342036-27342036 on Build GRCh38
rs754828619 Chr.16: 27342033-27342033 on Build GRCh38
rs758651293 Chr.16: 27342185-27342185 on Build GRCh38
rs55864075 Chr.16: 27342076-27342076 on Build GRCh38
rs373963941 Chr.16: 27342096-27342096 on Build GRCh38
rs201852059 Chr.16: 27342208-27342208 on Build GRCh38
rs765695378 Chr.16: 27342114-27342114 on Build GRCh38
rs1003024366 Chr.16: 27342044-27342044 on Build GRCh38
rs376483512 Chr.16: 27342086-27342086 on Build GRCh38
rs376961404 Chr.16: 27342075-27342075 on Build GRCh38
rs754863252 Chr.16: 27342070-27342070 on Build GRCh38
rs777645418 Chr.16: 27342165-27342165 on Build GRCh38
rs780462972 Chr.16: 27342192-27342192 on Build GRCh38
rs891889842 Chr.16: 27342052-27342052 on Build GRCh38
rs553282601 Chr.16: 27342126-27342126 on Build GRCh38
rs927581850 Chr.16: 27342218-27342218 on Build GRCh38
rs762532424 Chr.16: 27342128-27342128 on Build GRCh38
rs747063458 Chr.16: 27342196-27342196 on Build GRCh38
rs550734991 Chr.16: 27342093-27342093 on Build GRCh38
rs372988882 Chr.16: 27342153-27342153 on Build GRCh38
rs778990471 Chr.16: 27342085-27342085 on Build GRCh38
rs980433924 Chr.16: 27342098-27342098 on Build GRCh38
rs766039972 Chr.16: 27342134-27342134 on Build GRCh38
rs778084490 Chr.16: 27342072-27342072 on Build GRCh38
rs930075388 Chr.16: 27342182-27342182 on Build GRCh38
rs749110570 Chr.16: 27342177-27342177 on Build GRCh38
rs769011169 Chr.16: 27342095-27342095 on Build GRCh38
rs1021620944 Chr.16: 27342067-27342067 on Build GRCh38
rs370200363 Chr.16: 27342082-27342082 on Build GRCh38
rs200293983 Chr.16: 27342219-27342219 on Build GRCh38
rs751032016 Chr.16: 27342143-27342143 on Build GRCh38
rs1010238984 Chr.16: 27342062-27342062 on Build GRCh38

View Full Product Details