Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 109959239-109959510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGF
Entrez Gene ID
1950
Gene Name
epidermal growth factor
Gene Aliases
HOMG4, URG
UniGene
Hs.419815
Gene Chromosome Location
Chr.4: 109912883-110013079 on Build GRCh38
Transcripts
NM_001178130.2 NM_001178131.2 NM_001963.5 XM_005262796.2 XM_011531707.1 XM_017007845.1 XM_017007846.1 XM_017007847.1 XM_017007848.1 XM_017007849.1 XM_017007850.1 XM_017007851.1 XM_017007852.1 XM_017007853.1 XM_017007854.1 XM_017007855.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7163283 c.1027G>C p.E343Q Chr.4: 109959398-109959398 on Build GRCh38
2154679 c.1038C>A p.A346A Chr.4: 109959409-109959409 on Build GRCh38
1694814 c.1046G>A p.R349Q Chr.4: 109959417-109959417 on Build GRCh38
3006353 c.1036G>A p.A346T Chr.4: 109959407-109959407 on Build GRCh38
3006354 c.1052G>A p.R351Q Chr.4: 109959423-109959423 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762757772 Chr.4: 109959406-109959406 on Build GRCh38
rs981721153 Chr.4: 109959401-109959401 on Build GRCh38
rs778984329 Chr.4: 109959472-109959472 on Build GRCh38
rs546599803 Chr.4: 109959267-109959267 on Build GRCh38
rs996775239 Chr.4: 109959421-109959421 on Build GRCh38
rs146070944 Chr.4: 109959265-109959265 on Build GRCh38
rs772589488 Chr.4: 109959434-109959434 on Build GRCh38
rs745521297 Chr.4: 109959490-109959490 on Build GRCh38
rs747451818 Chr.4: 109959478-109959478 on Build GRCh38
rs764397759 Chr.4: 109959467-109959468 on Build GRCh38
rs756249353 Chr.4: 109959465-109959466 on Build GRCh38
rs776095220 Chr.4: 109959407-109959407 on Build GRCh38
rs986240376 Chr.4: 109959462-109959462 on Build GRCh38
rs145706760 Chr.4: 109959349-109959349 on Build GRCh38
rs962299408 Chr.4: 109959419-109959419 on Build GRCh38
rs368465628 Chr.4: 109959358-109959358 on Build GRCh38
rs566637639 Chr.4: 109959280-109959280 on Build GRCh38
rs766742641 Chr.4: 109959355-109959356 on Build GRCh38
rs200099199 Chr.4: 109959423-109959423 on Build GRCh38
rs147880662 Chr.4: 109959303-109959303 on Build GRCh38
rs756145526 Chr.4: 109959308-109959308 on Build GRCh38
rs536925744 Chr.4: 109959476-109959476 on Build GRCh38
rs539312498 Chr.4: 109959418-109959419 on Build GRCh38
rs761387138 Chr.4: 109959269-109959269 on Build GRCh38
rs1023773707 Chr.4: 109959366-109959366 on Build GRCh38
rs779402563 Chr.4: 109959354-109959354 on Build GRCh38
rs765024945 Chr.4: 109959274-109959274 on Build GRCh38
rs753111626 Chr.4: 109959452-109959453 on Build GRCh38
rs756200933 Chr.4: 109959446-109959446 on Build GRCh38
rs11568927 Chr.4: 109959477-109959477 on Build GRCh38
rs970434678 Chr.4: 109959373-109959373 on Build GRCh38
rs777994103 Chr.4: 109959315-109959315 on Build GRCh38
rs759220369 Chr.4: 109959425-109959425 on Build GRCh38
rs767856854 Chr.4: 109959423-109959446 on Build GRCh38
rs376075365 Chr.4: 109959475-109959475 on Build GRCh38
rs766127465 Chr.4: 109959278-109959278 on Build GRCh38
rs371001195 Chr.4: 109959277-109959277 on Build GRCh38
rs76435576 Chr.4: 109959447-109959447 on Build GRCh38
rs760103609 Chr.4: 109959422-109959423 on Build GRCh38
rs769369081 Chr.4: 109959391-109959391 on Build GRCh38
rs765930486 Chr.4: 109959408-109959408 on Build GRCh38
rs757558964 Chr.4: 109959454-109959454 on Build GRCh38
rs776112801 Chr.4: 109959374-109959374 on Build GRCh38
rs538492532 Chr.4: 109959285-109959285 on Build GRCh38
rs769277322 Chr.4: 109959489-109959489 on Build GRCh38
rs749456522 Chr.4: 109959323-109959323 on Build GRCh38
rs920813055 Chr.4: 109959438-109959438 on Build GRCh38
rs761442826 Chr.4: 109959388-109959388 on Build GRCh38
rs772688411 Chr.4: 109959371-109959371 on Build GRCh38
rs773001111 Chr.4: 109959400-109959400 on Build GRCh38
rs767519919 Chr.4: 109959431-109959431 on Build GRCh38

View Full Product Details