Gene Transcripts

Gene Symbol
Ppp1r26
Entrez Gene ID
241289
Gene Name
protein phosphatase 1, regulatory subunit 26
Gene Aliases
Gm347, Kiaa0649, mKIAA0649
Chromosome Location
Chr. 2: 28446155 - 28455508 on Build GRCm38
UniGene ID
Mm.330095
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005420.1 NP_001005420.1 3 1090
GenBank mRNA AK147361.1 - 1 3686
AK172993.1 - 1 848
BC070440.1 - 3 1090 30362439

View Full Product Details