Gene Transcripts

Gene Symbol
Kcna3
Entrez Gene ID
16491
Gene Name
potassium voltage-gated channel, shaker-related subfamily, member 3
Gene Aliases
Kca1-3, Kv1.3, Mk-3
Chromosome Location
Chr. 3: 107036162 - 107038129 on Build GRCm38
UniGene ID
Mm.30640
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008418.2 NP_032444.2 1 1281
GenBank mRNA BC132349.1 - 1 1118
BC137667.1 - 1 1118 8860686

View Full Product Details