Gene Transcripts

Gene Symbol
En2
Entrez Gene ID
13799
Gene Name
engrailed 2
Gene Aliases
BB131122, En-2
Chromosome Location
Chr. 5: 28165696 - 28172166 on Build GRCm38
UniGene ID
Mm.4298
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010134.3 NP_034264.1 2 1656
GenBank mRNA AK157766.1 - 2 1658
L12705.1 - 2 872

View Full Product Details