Gene Transcripts

Gene Symbol
Naa10
Entrez Gene ID
56292
Gene Name
N(alpha)-acetyltransferase 10, NatA catalytic subunit
Gene Aliases
2310039H09Rik, Ard1, Ard1a, Te2
Chromosome Location
Chr. X: 73916870 - 73921944 on Build GRCm38
UniGene ID
Mm.246654
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177965.1 NP_001171436.1 7 568
NM_019870.3 NP_063923.1 7 568
GenBank mRNA AJ627039.1 - 6 387
AK003806.1 - 7 505
AK009697.1 - 7 479
AK014469.1 - 7 497
AY183136.1 - 7 537
AY183137.1 - 7 537
BC027219.1 - 7 469 3673739

Gene Symbol
Arhgap4
Entrez Gene ID
171207
Gene Name
Rho GTPase activating protein 4
Gene Aliases
A130009C12Rik, A530015A20Rik, Rgc1, c1, mKIAA0131
Chromosome Location
Chr. X: 73894352 - 73911346 on Build GRCm38
UniGene ID
Mm.482337
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK129060.1 - 480

View Full Product Details