Gene Transcripts

Gene Symbol
Csnk1a1
Entrez Gene ID
113927
Gene Name
casein kinase 1, alpha 1
Gene Aliases
Chromosome Location
Chr. 18: 56887722 - 56910610 on Build Rnor_6.0
UniGene ID
Rn.23810
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053615.1 NP_446067.1 4, 5 451
GenBank mRNA BC127486.1 - 677
U77582.1 - 4, 5 451

View Full Product Details