Gene Transcripts

Gene Symbol
Mycbp2
Entrez Gene ID
290447
Gene Name
MYC binding protein 2, E3 ubiquitin protein ligase
Gene Aliases
Pam, Phr1
Chromosome Location
Chr. 15: 87404907 - 87540795 on Build Rnor_6.0
UniGene ID
Rn.13538
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106055.2 NP_001099525.2 52 9457
GenBank mRNA FQ232687.1 - 1 163

View Full Product Details