Gene Transcripts

Gene Symbol
Ifit2
Entrez Gene ID
294091
Gene Name
interferon-induced protein with tetratricopeptide repeats 2
Gene Aliases
Chromosome Location
Chr. 1: 252894663 - 252900727 on Build Rnor_6.0
UniGene ID
Rn.6175
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024753.1 NP_001019924.1 2 1108
GenBank mRNA BC097384.1 - 2 1108

View Full Product Details