Gene Transcripts

Gene Symbol
Olr830
Entrez Gene ID
405210
Gene Name
olfactory receptor 830
Gene Aliases
Chromosome Location
Chr. 4: 148664043 - 148665912 on Build Rnor_6.0
UniGene ID
Rn.143080
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000900.1 NP_001000900.1 1 578
XM_006237195.1 XP_006237257.1 2 649

View Full Product Details