Gene Transcripts

Gene Symbol
Amigo2
Entrez Gene ID
300186
Gene Name
adhesion molecule with Ig like domain 2
Gene Aliases
Ali1
Chromosome Location
Chr. 7: 138704124 - 138707221 on Build Rnor_6.0
UniGene ID
Rn.98123
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182816.2 NP_877968.2 1 2168
XM_006242290.1 XP_006242352.1 1 1533
GenBank mRNA AB078879.1 - 1 2168
AY237730.1 - 1 1472

View Full Product Details