Gene Transcripts

Gene Symbol
Thoc7
Entrez Gene ID
305714
Gene Name
THO complex 7 homolog (Drosophila)
Gene Aliases
RGD1305475
Chromosome Location
Chr. 15: 12569235 - 12584101 on Build Rnor_6.0
UniGene ID
Rn.22227
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271293.1 NP_001258222.1 2 133
NM_001271294.1 NP_001258223.1 2 177
XM_006251724.1 XP_006251786.1 2 257
XM_006251725.1 XP_006251787.1 2 144
GenBank mRNA EV771598.1 - 2 179
FM110383.1 - 2 93

View Full Product Details