Gene Transcripts

Gene Symbol
Scaf4
Entrez Gene ID
245924
Gene Name
SR-related CTD-associated factor 4
Gene Aliases
Sfrs15, Srsf15
Chromosome Location
Chr. 11: 30371710 - 30428175 on Build Rnor_6.0
UniGene ID
Rn.101886
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037347.2 NP_001032424.2 16 2566
GenBank mRNA U49058.1 - 15 2441

View Full Product Details