Gene Transcripts

Gene Symbol
Sp1
Entrez Gene ID
24790
Gene Name
Sp1 transcription factor
Gene Aliases
Chromosome Location
Chr. 7: 144014173 - 144044635 on Build Rnor_6.0
UniGene ID
Rn.44609
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012655.2 NP_036787.2 4 1842
GenBank mRNA D12768.1 - 4 1858

View Full Product Details