Gene Transcripts

Gene Symbol
Rcn3
Entrez Gene ID
494125
Gene Name
reticulocalbin 3, EF-hand calcium binding domain
Gene Aliases
Rem1, Rlp49
Chromosome Location
Chr. 1: 101076971 - 101086198 on Build Rnor_6.0
UniGene ID
Rn.2997
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008694.2 NP_001008694.1 3 391
GenBank mRNA BC083719.1 - 3 392
CK601329.1 - 3 391
FQ228883.1 - 3 411
L41683.1 - 2 106

View Full Product Details