Gene Transcripts

Gene Symbol
Poln
Entrez Gene ID
102555079
Gene Name
polymerase (DNA directed) nu
Gene Aliases
Chromosome Location
Chr. 14: 81837835 - 82037999 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006251412.1 XP_006251474.1 7, 8 1696
XM_006251414.2 XP_006251476.1 7, 8 1696
XM_008770382.1 XP_008768604.1 7, 8 1696
XM_008770383.1 XP_008768605.1 7, 8 1255
XM_008770384.1 XP_008768606.1 7, 8 1696

View Full Product Details