Gene Transcripts

Gene Symbol
PTEN
Entrez Gene ID
5728
Gene Name
phosphatase and tensin homolog
Gene Aliases
10q23del, BZS, CWS1, DEC, GLM2, MHAM, MMAC1, PTEN1, TEP1
Chromosome Location
Chr.10: 87863438 - 87971930 on Build GRCh38
Uni Gene
Hs.500466
Interrogated Sequence Translated Protein
RefSeq NM_000314.6 NP_000305.3
NM_001304717.2 NP_001291646.2
NM_001304718.1 NP_001291647.1
GenBank mRNA AA017584.1 -
AA836562.1 -
AI825848.1 -
AK021619.1 -
AK024986.1 -
AK124525.1 -
AK130076.1 -
AK313581.1 -
BC005821.2 -
BG772190.1 -
CA944942.1 -
CR450306.1 -
ENST00000371953.0 -
JF268690.1 -
KJ534922.1 -
U92436.1 -
U93051.1 -
U96180.1 -

View Full Product Details