Gene Transcripts

Gene Symbol
BMP2
Entrez Gene ID
650
Gene Name
bone morphogenetic protein 2
Gene Aliases
BDA2, BMP2A
Chromosome Location
Chr.20: 6767700 - 6780280 on Build GRCh38
Uni Gene
Hs.73853
Interrogated Sequence Translated Protein
RefSeq NM_001200.3 NP_001191.1
XM_011529323.2 XP_011527625.1
GenBank mRNA AA489383.1 -
BC069214.1 -
BM670141.1 -
ENST00000378827.0 -
KC294426.1 -
KF250425.1 -
M22489.1 -

View Full Product Details