Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_000314.6 | NP_000305.3 |
NM_001304717.2 | NP_001291646.2 | |
NM_001304718.1 | NP_001291647.1 | |
GenBank mRNA | AA017584.1 | - |
AA836562.1 | - | |
AI825848.1 | - | |
AK021619.1 | - | |
AK024986.1 | - | |
AK124525.1 | - | |
AK130076.1 | - | |
AK313581.1 | - | |
BC005821.2 | - | |
BG772190.1 | - | |
CA944942.1 | - | |
CR450306.1 | - | |
ENST00000371953.0 | - | |
JF268690.1 | - | |
KJ534922.1 | - | |
U92436.1 | - | |
U93051.1 | - | |
U96180.1 | - |