Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001178130.2 | NP_001171601.1 |
NM_001178131.2 | NP_001171602.1 | |
NM_001963.5 | NP_001954.2 | |
XM_005262796.2 | XP_005262853.1 | |
XM_011531707.1 | XP_011530009.1 | |
XM_017007845.1 | XP_016863334.1 | |
XM_017007846.1 | XP_016863335.1 | |
XM_017007847.1 | XP_016863336.1 | |
XM_017007848.1 | XP_016863337.1 | |
XM_017007849.1 | XP_016863338.1 | |
XM_017007850.1 | XP_016863339.1 | |
XM_017007851.1 | XP_016863340.1 | |
XM_017007852.1 | XP_016863341.1 | |
XM_017007853.1 | XP_016863342.1 | |
XM_017007854.1 | XP_016863343.1 | |
XM_017007855.1 | XP_016863344.1 | |
GenBank mRNA | AI628974.1 | - |
AK299306.1 | - | |
AK307734.1 | - | |
AY548762.1 | - | |
BC093731.1 | - | |
BC113461.1 | - | |
BC143357.1 | - | |
CA437074.1 | - | |
CU446298.1 | - | |
DB227969.1 | - | |
DB349864.1 | - | |
ENST00000265171.0 | - | |
JQ346088.1 | - | |
X04571.1 | - |