Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_002015.3 | NP_002006.2 |
XM_011535008.2 | XP_011533310.1 | |
XM_011535010.2 | XP_011533312.1 | |
GenBank mRNA | AF032885.1 | - |
AK310511.1 | - | |
BC021981.2 | - | |
BC070065.1 | - | |
BT007455.1 | - | |
BX648278.1 | - | |
ENST00000379561.0 | - | |
U02310.1 | - | |
U36922.1 | - |