Gene Transcripts

Gene Symbol
Bmp2
Entrez Gene ID
12156
Gene Name
bone morphogenetic protein 2
Gene Aliases
AI467020, Bmp2a
Chromosome Location
Chr.2: 133552159 - 133562896 on Build GRCm38
Uni Gene
Mm.103205
Interrogated Sequence Translated Protein
RefSeq NM_007553.3 NP_031579.2
GenBank mRNA AK132433.1 -
AK133923.1 -
AK142118.1 -
AK161862.1 -
AV333272.1 -
BC100344.1 -

View Full Product Details