Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_027470.3 | NP_081746.1 |
GenBank mRNA | AK017713.1 | - |
AK088512.1 | - | |
AK129298.2 | - | |
AK183660.1 | - | |
AY217016.1 | - | |
BB839524.1 | - | |
BC030389.1 | - | |
BC048238.1 | - | |
BQ572351.1 | - | |
BY197606.1 | - | |
CF172418.1 | - |