Gene Transcripts

Gene Symbol
Pten
Entrez Gene ID
19211
Gene Name
phosphatase and tensin homolog
Gene Aliases
2310035O07Rik, A130070J02Rik, AI463227, B430203M17Rik, MMAC1, TEP1
Chromosome Location
Chr.19: 32757577 - 32826160 on Build GRCm38
Uni Gene
Mm.245395
Interrogated Sequence Translated Protein
RefSeq NM_008960.2 NP_032986.1
GenBank mRNA AK009640.1 -
AK030750.1 -
AK037998.1 -
AK045001.1 -
AK051115.1 -
AK076980.1 -
AK080905.1 -
AK086551.1 -
AK087678.1 -
AK088717.1 -
AK138025.1 -
AK148736.1 -
AK164302.1 -
AK208703.1 -
AK212035.1 -
AK219196.1 -
BC021445.1 -
BC150968.1 -
BU701098.1 -
CK616227.1 -
U92437.1 -

View Full Product Details