Gene Transcripts

Gene Symbol
Foxo1
Entrez Gene ID
56458
Gene Name
forkhead box O1
Gene Aliases
AI876417, Afxh, FKHR, Fkhr1, Foxo1a
Chromosome Location
Chr.3: 52268337 - 52350109 on Build GRCm38
Uni Gene
Mm.29891
Interrogated Sequence Translated Protein
RefSeq NM_019739.3 NP_062713.2
GenBank mRNA AF114258.1 -
AF126056.1 -
AJ252157.1 -
AK029013.1 -
AK030304.1 -
AK039265.1 -
AK043583.1 -
AK053633.1 -
AK054289.1 -
AK137629.1 -
AK154041.1 -
AK185432.1 -
AK192846.1 -
AK211466.1 -
AK211487.1 -
AK212278.1 -
AK212379.1 -

View Full Product Details