Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_008960.2 | NP_032986.1 |
GenBank mRNA | AK009640.1 | - |
AK030750.1 | - | |
AK037998.1 | - | |
AK045001.1 | - | |
AK051115.1 | - | |
AK076980.1 | - | |
AK080905.1 | - | |
AK086551.1 | - | |
AK087678.1 | - | |
AK088717.1 | - | |
AK138025.1 | - | |
AK148736.1 | - | |
AK164302.1 | - | |
AK208703.1 | - | |
AK212035.1 | - | |
AK219196.1 | - | |
BC021445.1 | - | |
BC150968.1 | - | |
BU701098.1 | - | |
CK616227.1 | - | |
U92437.1 | - |