Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_019739.3 | NP_062713.2 |
GenBank mRNA | AF114258.1 | - |
AF126056.1 | - | |
AJ252157.1 | - | |
AK029013.1 | - | |
AK030304.1 | - | |
AK039265.1 | - | |
AK043583.1 | - | |
AK053633.1 | - | |
AK054289.1 | - | |
AK137629.1 | - | |
AK154041.1 | - | |
AK185432.1 | - | |
AK192846.1 | - | |
AK211466.1 | - | |
AK211487.1 | - | |
AK212278.1 | - | |
AK212379.1 | - |