Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_009096.3 | NP_033122.1 |
GenBank mRNA | AK012284.1 | - |
AK012828.1 | - | |
AK013333.1 | - | |
AK013825.1 | - | |
AK028205.1 | - | |
AK050609.1 | - | |
AK146952.1 | - | |
AK161991.1 | - | |
AK166680.1 | - | |
AK166727.1 | - | |
AK168050.1 | - | |
AK168496.1 | - | |
AK169624.1 | - | |
BC010604.1 | - | |
BC090392.1 | - | |
BC092050.1 | - | |
Y00348.1 | - |