Gene Transcripts
- Gene Symbol
- 4732416N19Rik
- Entrez Gene ID
- 320737
- Gene Name
- RIKEN cDNA 4732416N19 gene
- Gene Aliases
- Chromosome Location
- Chr. 6: 148212288 - 148235663 on Build GRCm38
- UniGene ID
- Mm.137858
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_015615.1 |
|
2 |
375 |
|
GenBank mRNA |
AK005757.1 |
- |
2 |
375 |
|
AK033816.1 |
- |
|
534 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY705917.1 |
- |
2 |
375 |
|
Chr. 6 - 148212288 - 148235666 [+] |
View Full Product Details