Gene Transcripts
- Gene Symbol
- FLJ37453
- Entrez Gene ID
- 729614
- Gene Name
- uncharacterized LOC729614
- Gene Aliases
- Chromosome Location
- Chr. 1: 15834215 - 15848147 on Build GRCh38
- UniGene ID
- Hs.119298
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024279.1 |
|
2 |
2205 |
|
GenBank mRNA |
AK094772.1 |
- |
1 |
1119 |
|
AK124018.1 |
- |
2 |
2206 |
|
View Full Product Details