Gene Transcripts
- Gene Symbol
- LOC100303749
- Entrez Gene ID
- 100303749
- Gene Name
- LSM3 homolog, U6 small nuclear RNA associated (S. cerevisiae) pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 5: 65944805 - 65945573 on Build GRCh38
- UniGene ID
- Hs.608060
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028474.1 |
|
1 |
205 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BU568479.1 |
- |
|
369 |
|
Chr. - - [-] |
View Full Product Details