Gene Transcripts

Gene Symbol
GEN1
Entrez Gene ID
348654
Gene Name
GEN1, Holliday junction 5' flap endonuclease
Gene Aliases
Gen
Chromosome Location
Chr. 2: 17753687 - 17785920 on Build GRCh38
UniGene ID
Hs.467793
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK093631.1 - 1 872
BC038243.1 - 1 1136


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CR617603.1 - 1 1735 Chr. 2 - 17753296 - 17755047 [+]

View Full Product Details