Gene Transcripts
- Gene Symbol
- LOC101927829
- Entrez Gene ID
- 101927829
- Gene Name
- uncharacterized LOC101927829
- Gene Aliases
- Chromosome Location
- Chr. 3: 20342468 - 20350928 on Build GRCh38
- UniGene ID
- Hs.639381
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110814.1 |
|
1 |
416 |
|
GenBank mRNA |
BC041892.1 |
- |
1 |
416 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000455626.1 |
- |
1 |
416 |
|
Chr. 3 - 20342468 - 20350928 [+] |
View Full Product Details