Gene Transcripts
- Gene Symbol
- 4933407O12Rik
- Entrez Gene ID
- 71065
- Gene Name
- RIKEN cDNA 4933407O12 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.426426
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK016733.1 |
- |
1 |
968 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX520778.1 |
- |
1 |
273 |
|
Chr. 17 - 80063496 - 80064025 [+] |
View Full Product Details