Gene Transcripts
- Gene Symbol
- 4930473O22Rik
- Entrez Gene ID
- 75025
- Gene Name
- RIKEN cDNA 4930473O22 gene
- Gene Aliases
- Chromosome Location
- Chr. 10: 97590984 - 97596931 on Build GRCm38
- UniGene ID
- Mm.159401
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045356.1 |
|
1 |
148 |
|
GenBank mRNA |
AK015568.1 |
- |
1 |
150 |
|
View Full Product Details