Gene Transcripts
- Gene Symbol
- 6720469O03Rik
- Entrez Gene ID
- 77738
- Gene Name
- RIKEN cDNA 6720469O03 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020153.1 |
- |
1 |
42 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY719411.1 |
- |
1 |
42 |
|
Chr. 7 - 37365441 - 37366097 [-] |
View Full Product Details