Gene Transcripts
- Gene Symbol
- LOC497255
- Entrez Gene ID
- 497255
- Gene Name
- uncharacterized LOC497255
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.23084
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK078348.1 |
- |
1 |
231 |
|
View Full Product Details