Gene Transcripts
- Gene Symbol
- 4930511A08Rik
- Entrez Gene ID
- 78178
- Gene Name
- RIKEN cDNA 4930511A08 gene
- Gene Aliases
- Chromosome Location
- Chr. 6: 94132922 - 94246031 on Build GRCm38
- UniGene ID
- Mm.452749
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK019673.1 |
- |
|
256 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY715287.1 |
- |
3 |
256 |
|
Chr. 6 - 94132922 - 94283025 [-] |
View Full Product Details