Gene Transcripts
- Gene Symbol
- Gm11837
- Entrez Gene ID
- 100038514
- Gene Name
- predicted gene 11837
- Gene Aliases
- OTTMUSG00000004551
- Chromosome Location
- Chr. 4: 14929829 - 14953031 on Build GRCm38
- UniGene ID
- Mm.331495
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK005641.1 |
- |
6 |
661 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CJ189634.1 |
- |
2 |
454 |
|
Chr. 4 - 14947479 - 14953026 [+] |
View Full Product Details