Gene Transcripts

Gene Symbol
Gm11837
Entrez Gene ID
100038514
Gene Name
predicted gene 11837
Gene Aliases
OTTMUSG00000004551
Chromosome Location
Chr. 4: 14929829 - 14953031 on Build GRCm38
UniGene ID
Mm.331495
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK005641.1 - 6 399


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BU962870.1 - 6 572 Chr. 4 - 14930683 - 14952917 [+]
GenBankmRNA CA463133.1 - 6 584 Chr. 4 - 14930669 - 14952832 [+]
GenBankmRNA CJ189634.1 - 2 192 Chr. 4 - 14947479 - 14953026 [+]
GenBankmRNA CA464091.1 - 6 592 Chr. 4 - 14930661 - 14952967 [+]
GenBankmRNA CD363818.1 - 6 613 Chr. 4 - 14929908 - 14952893 [+]
GenBankmRNA CA463179.1 - 6 580 Chr. 4 - 14930676 - 14952890 [+]
GenBankmRNA BU938202.1 - 460 Chr. - - [-]

View Full Product Details