Gene Transcripts
- Gene Symbol
- Gm13799
- Entrez Gene ID
- 102636638
- Gene Name
- predicted gene 13799
- Gene Aliases
- OTTMUSG00000014516
- Chromosome Location
- Chr. 2: 95838088 - 95840594 on Build GRCm38
- UniGene ID
- Mm.399115
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK029614.1 |
- |
3 |
302 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AV260572.2 |
- |
3 |
432 |
|
Chr. 2 - 95838099 - 95840591 [+] |
GenBankmRNA |
BB615000.1 |
- |
3 |
302 |
|
Chr. 2 - 95838088 - 95840554 [+] |
View Full Product Details