Gene Transcripts
- Gene Symbol
- 1700086P04Rik
- Entrez Gene ID
- 67349
- Gene Name
- RIKEN cDNA 1700086P04 gene
- Gene Aliases
- Chromosome Location
- Chr. 4: 129050087 - 129057996 on Build GRCm38
- UniGene ID
- Mm.174885
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK005980.1 |
- |
1 |
108 |
|
AK007024.1 |
- |
1 |
100 |
|
AK019743.1 |
- |
1 |
108 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY707078.1 |
- |
1 |
100 |
|
Chr. 4 - 129050126 - 129057974 [-] |
GenBankmRNA |
BY715641.1 |
- |
1 |
104 |
|
Chr. 4 - 129050625 - 129057980 [-] |
GenBankmRNA |
BY706111.1 |
- |
1 |
104 |
|
Chr. 4 - 129050421 - 129057980 [-] |
View Full Product Details