Gene Transcripts
- Gene Symbol
- LOC101928710
- Entrez Gene ID
- 101928710
- Gene Name
- uncharacterized LOC101928710
- Gene Aliases
- Chromosome Location
- Chr. 17: 78617384 - 78632057 on Build GRCh38
- UniGene ID
- Hs.464224
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110848.1 |
|
5 |
2051 |
|
NR_110849.1 |
|
5 |
742 |
|
GenBank mRNA |
BG613706.1 |
- |
5 |
744 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000590023.5 |
- |
3 |
296 |
|
Chr. 17 - 78617389 - 78630170 [-] |
GenBankmRNA |
BG203385.1 |
- |
2 |
160 |
|
Chr. 17 - 78617388 - 78624318 [-] |
View Full Product Details