Gene Transcripts
- Gene Symbol
- 7330404K18Rik
- Entrez Gene ID
- 78145
- Gene Name
- RIKEN cDNA 7330404K18 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020178.1 |
- |
2 |
432 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY719572.1 |
- |
2 |
432 |
|
Chr. 14 - 20398359 - 20398930 [+] |
View Full Product Details