Gene Transcripts
- Gene Symbol
- Ccdc34os
- Entrez Gene ID
- 75517
- Gene Name
- coiled-coil domain containing 34, opposite strand
- Gene Aliases
- 1700024P03Rik
- Chromosome Location
- Chr. 2: 110031250 - 110050479 on Build GRCm38
- UniGene ID
- Mm.363927
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040508.1 |
|
3, 4 |
530 |
|
GenBank mRNA |
AK006310.1 |
- |
3, 4 |
531 |
|
View Full Product Details