Gene Transcripts

Gene Symbol
Fbxo11
Entrez Gene ID
225055
Gene Name
F-box protein 11
Gene Aliases
C80048, Jf
Chromosome Location
Chr. 17: 87990859 - 88065285 on Build GRCm38
UniGene ID
Mm.386857
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081034.1 NP_001074503.1 11 1400
GenBank mRNA BC055343.1 - 2 142
BC110686.1 - 11 1139
BC117884.1 - 11 1146 40096054
BC117885.1 - 11 1150 40096055
BC128479.1 - 11 1150 40096053

View Full Product Details