Gene Transcripts
- Gene Symbol
- Olfr774
- Entrez Gene ID
- 258232
- Gene Name
- olfactory receptor 774
- Gene Aliases
- MOR111-11, MOR111-2
- Chromosome Location
- Chr. 10: 129238151 - 129239089 on Build GRCm38
- UniGene ID
- Mm.391464
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207620.1 |
NP_997503.1
|
1 |
240 |
|
- Gene Symbol
- Olfr776
- Entrez Gene ID
- 404321
- Gene Name
- olfactory receptor 776
- Gene Aliases
- MOR111-12
- Chromosome Location
- Chr. 10: 129260963 - 129261901 on Build GRCm38
- UniGene ID
- Mm.355212
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207559.1 |
NP_997442.1
|
1 |
240 |
|
- Gene Symbol
- Olfr787
- Entrez Gene ID
- 258069
- Gene Name
- olfactory receptor 787
- Gene Aliases
- MOR111-10
- Chromosome Location
- Chr. 10: 129461940 - 129463616 on Build GRCm38
- UniGene ID
- Mm.376120
- Species
- Mus musculus
View Full Product Details