Gene Transcripts

Gene Symbol
Snrpg
Entrez Gene ID
68011
Gene Name
small nuclear ribonucleoprotein polypeptide G
Gene Aliases
2810024K17Rik, AL022803, SMG, sm-G, snRNP-G
Chromosome Location
Chr. 6: 86371540 - 86378902 on Build GRCm38
UniGene ID
Mm.276802
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026506.2 NP_080782.1 4 364
GenBank mRNA AK011129.1 - 4 363
AK012802.1 - 4 353
AK013235.1 - 4 365
AK017852.1 - 4 713
BC027499.1 - 4 337 1430048
BC051470.1 - 4 409 1244608
BC094411.1 - 4 351 1380524

View Full Product Details