Gene Transcripts

Gene Symbol
Hspb1
Entrez Gene ID
24471
Gene Name
heat shock protein B1
Gene Aliases
Hsp25, Hsp27
Chromosome Location
Chr. 12: 23839420 - 23840981 on Build Rnor_6.0
UniGene ID
Rn.3841
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA FQ215417.1 - 618
FQ221491.1 - 3 618
FQ222013.1 - 3 618
FQ222462.1 - 3 618
M86389.1 - 3 599

View Full Product Details