Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031803.1 | NP_113991.1 | 9 | 983 | |
GenBank mRNA | AF059273.1 | - | 9 | 983 | |
AF205778.1 | - | 877 | |||
AF205779.1 | - | 877 | |||
AF205780.1 | - | 877 | |||
FQ211504.1 | - | 1027 |